Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water, ACS Reagent Grade, ASTM Type I, Ricca Chemical
SDP

Catalog No. R9150000500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
55 gal.
Packaging:
Cubitainer
Hard Pack
Natural Poly Bottle
Poly Drum
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R9150000500 500 mL Natural Poly Bottle
9150-32 1 L Natural Poly Bottle
9150-1 4 L Natural Poly Bottle
9150-25 10 L Cubitainer
9150-5 20 L Cubitainer
9150-5HP 20 L Hard Pack
13-622-621 55 gal. Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. R9150000500 Supplier Ricca Chemical Company Supplier No. R9150000500A
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
PubChem CID 962
ChEBI CHEBI:15377
SMILES O

Specifications

Color Colorless
Melting Point 0°C
CAS Min % 98.0
Boiling Point 100°C
CAS Max % 100.0
Conductivity max. 0.056
Synonym dihydrogen oxide
Solubility Information Miscible
IUPAC Name water
Molecular Weight (g/mol) 18.02
Formula Weight 18.01
Percent Purity 100%
Grade ACS
Packaging Natural Poly Bottle
Sterility Non-sterile
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Water, ASTM Type I
Show More Show Less
ShelfLife 18 months
DOTInformation Not Regulated by DOT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.