Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water, ACS Reagent, meets ASTM Type II specifications, Solstice

Catalog No. 6003086 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
5 gal.
4 x 4 L
Packaging:
Fiberboard Box
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
600-30-86 4 x 4 L Poly Bottle
60-030-87 5 gal. Fiberboard Box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 600-30-86 Supplier Solstice Supplier No. LP3664P
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Boiling Point 100°C
CAS Max % 100.0000%
Beilstein 2050024
Formula Weight 18
Grade ACS Reagent
Packaging Poly Bottle
Density 0.9971 g/cm3
Quantity 4 x 4 L
Chemical Name or Material Water, ACS Reagent, meets ASTM Type II specifications
ShelfLife 547 days from date of manufacture
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.