Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Water, CHROMASOLV™, for HPLC, Honeywell Riedel-de Haën™
Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
1 L
2.5 L
4 L
4 x 4 L
6 x 1 L
Packaging:
Glass Bottle
Description
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Specifications
Specifications
| Absorbance | max. 0.005 at 400 nm, max. 0.005 at 250 nm, max. 0.01 at 210 nm, max. 0.01 at 205 nm |
| Boiling Point | 100°C |
| CAS Max % | 100.0000% |
| Linear Formula | H2O |
| Beilstein | 2050024 |
| Formula Weight | 18.02 |
| Grade | HPLC |
| Packaging | Glass Bottle |
| Density | 1.000 g/mL at 3.98 °C(lit.) |
| Quantity | 2.5 L |
| Show More |
Safety and Handling
| ShelfLife | 720 days from date of manufacture |
| EINECSNumber | 231-791-2 |
| Recommended Storage | Room Temp |
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction