Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Science First™ Water Cycle Lab



Click to view available options
:
Water Cycle Lab
Description
Includes
- 16 thermometers
- Eight aluminum cans
- 16 sample containers
- Eight plastic plates
- Sponge
- Spoon
- Teacher pages
- Student instruction pages
- Student journal pages
Order Info
Grades K to 8; Class size: 32 students in groups of four
Specifications
Specifications
| Includes | 16 Thermometers, eight Aluminum cans, 16 sample containers, eight Plastic plates, sponge, spoon, teacher pages student instruction pages, student journal pages |
| Product Type | Water Cycle Lab |
| Grade | K to 8 |
| For Use With (Application) | Helps teach all parts of the water cycle of solid, liquid and gas |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction