Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water, Deionized, ASTM Type I, Certified ACS Grade, LabChem™

Catalog No. LC26740200
Change view
Click to view available options
Quantity:
1 L
4 L
10 L
20 L
200 L
Packaging:
Cubitainer
Poly Bottle
Poly Drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC26740200 200 L Poly Drum
LC267402 1 L Poly Bottle
LC267404 4 L Poly Bottle
LC267406 10 L Cubitainer
LC267405 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. LC26740200 Supplier LabChem, Inc. Supplier No. LC26740200
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym water, dihydrogen oxide, distilled water, purified water, water vapor, sterile water, water, purified, water, deionized, water, mineral, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Color Colorless
Melting Point 0°C
Boiling Point 100°C
CAS Max % 100
Conductivity ≤ 0.056uS/cm
Linear Formula H2O
Solubility Information Soluble in water
Formula Weight 18.02
Grade Certified ACS
Packaging Poly Drum
Viscosity 1.004cSt
Density 0.99823g/mL
Quantity 200 L
Physical Form Liquid
Chemical Name or Material Water, Deionized, ASTM Type I
Show More Show Less
Hazard Statement GHS H Statement
Product is not hazardous.
EINECSNumber 231-791-2
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.