Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water, GC Headspace Grade, Thermo Scientific™

Catalog No. w101 Shop All Fisherbrand Products
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle

Water processed to high purity for accurate and repeatable determination of trace levels of residual solvents

  • Every lot is tested to ensure interference free baselines for interference free determination of Class 1, 2, and 3 residual solvents
  • 1 ppm max. of Class 1 Solvents, 10 ppm max. of Class 2 solvents, and 50 ppm max of Class 3 residual solvents
  • This product is UV Absorbance Tested to ensure low organic contamination
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Absorbance 0.005 AU Maximum (400 nm), 0.005 AU Maximum (300 nm), 0.005 AU Maximum (280 nm), 0.005 AU Maximum (250 nm), 0.010 AU Maximum (220 nm), 0.010 AU Maximum (190 nm)
Formula Weight 18.01
Grade GC Headspace
Packaging Glass Bottle
Residue after Evaporation 1 ppm max
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Water
Recommended Storage Room temperature, packaged under inert atmosphere.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.