Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water Hardness Buffer, with Magnesium EDTA, Ricca Chemical
SDP

Catalog No. p-2917153
Click to view available options
Volume (Metric):
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle

APHA/ASTM/EPA for water hardness testing.

  • For water hardness testing.

Specifications

pH Range 7 to 14
Volume (Metric) 500 mL
Packaging Natural Poly Bottle
Quantity 500 mL
Sterility Non-sterile
Test Method APHA/ASTM/EPA for Water Hardness
Type Water Hardness Buffer
Volume (English) 16.9 oz.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.