Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Water Hardness Buffer with Magnesium EDTA, Reagents
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Specifications
Specifications
Color | Colorless |
Volume (Metric) | 20 L |
For Use With (Application) | Water Analysis |
Packaging | Cubitainer |
Quantity | 20 L |
Sterility | Non-sterile |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction