Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Water Hardness Indicator, 0.5% (w/v) Eriochrome Black T in 2-Methoxyethanol, Ricca Chemical

APHA/EPA for Water Hardness Testing
$47.10 - $192.80
Specifications
pH Range | Alkaline |
---|---|
Packaging | Amber Glass Bottle |
Type | Water Hardness Indicator |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
9300-4
![]() |
Ricca Chemical Company
93004 |
120 mL |
Each for $47.10
|
|
|||||
9300-16
![]() |
Ricca Chemical Company
930016 |
500 mL |
Each for $83.30
|
|
|||||
930032
![]() |
Ricca Chemical Company
930032 |
1 L |
Each for $192.80
|
|
|||||
Specifications
Alkaline | |
Amber Glass Bottle | |
Water Hardness Indicator |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title