Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Water Hardness Indicator 1% Eriochrome Black T in 20% Triethanolamine/80% Isopropanol, Reagents
Click to view available options
Quantity:
120 mL
500 mL
Specifications
Specifications
Color | Dark Red |
Volume (Metric) | 500 mL |
For Use With (Application) | Water Analysis |
Packaging | Glass Bottle |
Quantity | 500 mL |
Sterility | Non-sterile |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction