Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water Hardness Titrating Solution, Reagents

Catalog No. 01333385
Click to view available options
Quantity:
1 L
4 L
10 L
20 L

Water Hardness Titrating Solution 1 mL = 1 mg CaCO3, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Color Colorless
Volume (Metric) 20 L
For Use With (Application) Water Analysis
Packaging Cubitainer
Quantity 20 L
Sterility Non-sterile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.