Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rinse Solution, High Purity Water for Rinsing Electrodes and other Laboratory Instruments, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
500 mL
20 x 20 mL
200 x 20 mL
Packaging:
Natural Poly Bottle
Pouches
Specifications
Specifications
CAS Min % | 98.0 |
CAS Max % | 100.0 |
Percent Purity | 100% |
Grade | Laboratory |
Packaging | Pouches |
Quantity | 200 x 20 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction