Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Water (HPLC), Fisher Chemical PROMO
Shop All Fisherbrand Products
Click to view available options
:
4 L
Specifications
Specifications
| pH | 7 |
| Color | Colorless |
| Melting Point | 0°C |
| Boiling Point | 100°C |
| Formula Weight | 18.02 amu |
| Grade | Mobile Phase for HPLC Applications |
| Packaging | Glass Bottle |
| Residue after Evaporation | 1 ppm Max |
| Vapor Pressure | 17.5 mmHg at 20°C |
| Filtered Through | 0.2 Micron Filtered |
| Show More |
Safety and Handling
| Hazard Statement | Low hazard for usual industrial or commercial handling. |
| Precautionary Statement | Emergency Overview: Handle in accordance with good industrial hygiene and safety practice. Flush eyes with water as a precaution. Move to fresh air. If breathing is difficult, give oxygen. Do not induce vomiting. Obtain medical attention. NFPA Health:0 Flammability:0 Instability:0 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction