Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water, Molecular Biology Grade, Fisher BioReagents
GSA_VA

Catalog No. p~3795372 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 mL
1 L
4 L
10 L
20 L
Packaging:
Nalgene Poly Bottle
Poly Bottle
Poly CUBE
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP2819100 100 mL Poly Bottle
BP28191 1 L Nalgene Poly Bottle
BP28194 4 L Poly CUBE
BP281910 10 L Poly CUBE
BP281920 20 L Poly CUBE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. BP2819100
Only null left
Add to Cart

For many fundamental molecular biology procedures

Ultrapure, deionized, molecular biology grade water is ideal for many fundamental procedures such as PCR, electrophoresis, DNA sequencing, and buffers for enzymatic analyses.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

pH 7
Color Colorless
Melting Point 0°C
Boiling Point 100°C
Conductivity <2μS/cm
DNase DNase free
Merck Index 15, 10237
Formula Weight 18.02
Grade DNase-, RNase- and Protease-Free
Packaging Poly Bottle
Vapor Pressure 17.5mmHg at 20°C
Filtered Through Filtered through a 0.03μ filter
Treatment(s) Deionized
Density 1.000g/cm³
Quantity 100 mL
Physical Form Liquid
Chemical Name or Material Water, 0.03μm filtered to ensure high purity
Show More Show Less
Hazard Statement Low hazard for usual industrial or commercial handling.
Precautionary Statement Emergency Overview
Handle in accordance with good industrial hygiene and safety practice. Flush eyes with water as a precaution. Move to fresh air. If breathing is difficult, give oxygen. Do not induce vomiting. Obtain medical attention. . . . .

NFPA

Health:0
Flammability:0
Instability:0
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.