Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water, Optima™ for HPLC, Fisher Chemical™

Catalog No. p~216435 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Amber Soda Lime Glass 
Safe-Cote™ Amber Glass 
Wide Neck Amber Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
W71 1 L Wide Neck Amber Bottle
W7-4 4 L Amber Soda Lime Glass 
W7SK-4 4 L Safe-Cote™ Amber Glass 
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. W71
Only null left
Add to Cart

Improved filtration! High purity mobile phase for HPLC

  • Screened UV-absorbing contaminants to reduce interferences for smoother baselines
  • Tested for HPLC gradient suitability
  • Mobile phase for analytical and preparative separations
  • Now filters through 0.2 μm filter to remove particulates.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
MDL Number MFCD00011332
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Absorbance 0.005 AU max. at 254nm, 0.005 AU max. at 300nm, 0.005 AU max. at 400nm, 0.01 AU max. at 200nm
pH 7
Color Colorless
Melting Point 0°C
Boiling Point 100°C
Formula Weight 18.02
Grade High purity mobile phase for HPLC
Packaging Wide Neck Amber Bottle
Residue after Evaporation 1 ppm max.
Vapor Pressure 17.5mmHg at 20°C
Density 1000 g/cm³
Certificate of Analysis Certificate of Lot Analysis included with each bottle; UV and other chromatograms available on request.
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Water
Show More Show Less
Hazard Statement Low hazard for usual industrial or commercial handling.
Precautionary Statement Emergency Overview
Handle in accordance with good industrial hygiene and safety practice. Flush eyes with water as a precaution. Move to fresh air. If breathing is difficult, give oxygen. Do not induce vomiting. Obtain medical attention. . . . .

NFPA

Health:0
Flammability:0
Instability:0
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.