Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water, Optima™ LC/MS Grade, Fisher Chemical™ PROMO

Catalog No. W64FB25 Shop All Fisherbrand Products
Click to view available options
:
4 L

Glass bottles have fluorinated lining.

  • Water, H2O
  • CAS Number: 7732-18-5
  • Dihydrogen monoxide, dihydrogen oxide
  • 1 L
  • 0.005 AU at 280 nm
  • 0.005 AU at 260 nm
  • 0.005 AU at 254 nm
  • 0.01 AU at 210 nm
  • 0.01 AU at 230 nm
  • 0.01 AU at 240 nm
  • 0.01 AU at 220 nm
  • 100 deg.C, 20 ppb max
  • 18.015 g/mol
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015 g/mol
MDL Number 11332
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

Absorbance ≤0.005 AU at 254 nm, ≤0.005 AU at 260 nm, ≤0.005 AU at 280 nm, ≤0.01 AU at 210 nm, ≤0.01 AU at 220 nm, ≤0.01 AU at 230 nm, ≤0.01 AU at 240 nm
pH 7
Color Colorless
Melting Point 0°C
Boiling Point 100°C
Merck Index 15, 10237
Formula Weight 18.02 amu
Grade LC-MS and UHPLC-UV
Packaging Glass Bottle with Fluorinated Lining
Residue after Evaporation ≤1ppm
Vapor Pressure 17.5 mmHg at 20°C
Filtered Through 0.03 Micron Filtered
Density 1.000 g/cm3
Certificate of Analysis Actual Lot Analysis reported on label; Certificate of Actual Lot Analysis available on request
Quantity 4 L
Physical Form Liquid
Chemical Name or Material Water
Show More Show Less
Hazard Statement Low hazard for usual industrial or commercial handling.
Precautionary Statement

Emergency Overview: Handle in accordance with good industrial hygiene and safety practice. Flush eyes with water as a precaution. Move to fresh air. If breathing is difficult, give oxygen. Do not induce vomiting. Obtain medical attention.

NFPA

Health: 0

Flammability: 0

Instability: 0

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.