Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Water, for qPCR or RT-qPCR, Fisher BioReagents
Shop All Fisherbrand Products
Click to view available options
Quantity:
1 L
10 L
Packaging:
Poly Bottle
Poly CUBE
Description
- Minimize the risk of false positive signals
- Tested for DNase, RNase, protease, nickase, and E. coli and human genomic DNA using sensitive assays
- Evaluated for metal impurities at low ppb concentrations to ensure optimal enzymatic activity, especially for polymerase during PCR
- Sub-micron filtered to remove particulates
- Suitable for use in qPCR and RT-qPCR for genomics work, microarrays, and other sensitive life science applications
- Also useful for the manufacturing of PCR-related kits and mastermixes

Specifications
Specifications
pH | 5.4 to 7.0 |
Color | Clear, Colorless |
Melting Point | 0°C |
Boiling Point | 100°C |
Conductivity | ≤2μS/cm |
DNase | Not detected |
Endotoxin | <0.1EU/mL |
Formula Weight | 18.02 |
Grade | qPCR |
Packaging | Poly CUBE |
Show More |
Safety and Handling
Recommended Storage | 4°C; Ships at ambient temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction