Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water, Reagent, ACS, Spectrum™ Chemical
SDP

Catalog No. 18607733 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
1 L
20 L
4 x 4 L
6 x 1 L
200 L
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-733 20 L Poly Bottle
18-607-729 1 L Poly Bottle
18-607-731 6 x 1 L Poly Bottle
18-607-732 4 x 4 L Poly Bottle
18-607-734 200 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-607-733 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. W100020LTCU
Add to Cart
Add to Cart

W1000, 7732-18-5, H2O

Spectrum™ Chemical Water, Reagent, ACS is used in many chemical reactions. Due to its ability to dissolve many ionic compounds, water is a common solvent in inorganic reactions. It is, however, rarely used as a reaction solvent since it is amphoteric and is not able to dissolve reactants very well. As an ACS grade Reagent, Spectrum Chemical manufactured water is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

CAS Max % 1
Grade ACS
Packaging Poly Bottle
Quantity 20 L
Chemical Name or Material Water
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.