Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Water Rocket and Class Packs

Click to view available options
Product Type:
Water Rocket Class Pack
Water Rocket Single Kit
Includes:
1 Molded nozzle, 1 die cut fin set, 1 soft Foam bumper, assembly instructions
25 Precut rigid Foam fin set, 25 translucent molded nozzles, 25 Foam recovery bumpers, color assembly and use instructions
Description
Requires
Standard bicycle pump with a pressure gauge; assembly of foam fins requires low temperature hot glue gun
Packs consist of:
- Pre-cut rigid foam fin sets
- Translucent molded nozzles
- Foam recovery bumpers
- Color assembly and use instructions
Specifications
Specifications
Product Type | Water Rocket Single Kit |
Includes | 1 Molded nozzle, 1 die cut fin set, 1 soft Foam bumper, assembly instructions |
For Use With (Application) | Perfect low cost flying rocket without the hassles of traditional solid propellant rockets! Theres no fire danger (the rocket fuel is water) and no HazMat fees to ship them! |
Subject | Physics |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction