Learn More
Water Solution, 0.05% (w/v) Ammonium Formate, 0.1% (v/v) formic acid, Solstice
Shop All Solstice Research Chemicals ProductsDescription
- High purity chromatography solvents
- Suitability tested for specified use from HPLC up to UHPLC-MS applications.
Specifications
Specifications
| Density | 0.9972g/mL at 25°C |
| Quantity | 4 x 4 L |
| Packaging | Glass Bottle |
| Linear Formula | H2O |
| UN Number | NONH for all modes of transport |
| Chemical Name or Material | Water solution, Contains0.05% (w/v) ammonium formate, 0.1% (v/v) formic acid |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.