Learn More
BINDER™ Water Supply Set
Shop All BINDER Inc. Products
Click to view available options
Model:
Free-standing
Rear-mounting
Description
- External
- Includes fresh and waste water containers (20 L), cabling, and pump
Specifications
Specifications
Description | Water supply set - external, for hanging from the back of the device, consisting of fresh- and waste-water containers (20 liters each), cabling and pump |
For Use With (Equipment) | BINDER Chambers Series KBWF, KBF, KBF P, KBF LQC, KMF |
Material | Plastic |
Type | Water Supply |
Includes | Fresh- and Waste Water Containers (20 Liters Each), Cabling and Pump |
Model | Rear-mounting |