Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water, Ultra Trace Elemental Analysis Grade, Fisher Chemical™

Catalog No. p~216448 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2 L
Packaging:
LDPE Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
W9-500 500 mL LDPE Bottle
W9-1 1 L LDPE Bottle
W92 2 L LDPE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. W9-500
Only null left
Add to Cart

Packaged in specially precleaned LDPE bottles to protect product integrity

  • Evaluated for 65 metal impurities in ppt range
  • Also specified for ≤1ppb for chloride, phosphate and sulfate anion impurities
  • Packaged in precleaned LDPE bottles to maintain product integrity
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.015
MDL Number 11332
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name oxidane
SMILES O

Specifications

pH 7
Color Colorless
Melting Point 0°C
Boiling Point 100°C
Formula Weight 18.02
Grade Ultra Trace Elemental Analysis
Packaging LDPE Bottle
Vapor Pressure 17.5mmHg at 20°C
Density 1.000g/cm³
Certificate of Analysis Certificate of Lot Analysis included with each bottle.
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Water
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.