Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water, USP/EP/JP Purified, Ricca Chemical
SDP

Catalog No. r918900020f Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
20 L
Packaging:
Cubitainer
Hard Pack

Catalog No. R918900020F

Supplier: Ricca Chemical Company R918900020F

Only null left
Add to Cart
Add to Cart

USP/EP Purified Water

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILES O

Specifications

pH 5 to 7
Color Colorless
CAS Max % 100
Conductivity max. 1 μS/cm
Endotoxin max. 0.25 EU/mL at time of manufacture
IUPAC Name water
Molecular Weight (g/mol) 18.02
Percent Purity 100%
Grade EP/JP/USP
Packaging Cubitainer
Residue after Evaporation max. 0.001%
Sterility Non-sterile
Quantity 20 L
Chemical Name or Material Water
Show More Show Less
ShelfLife 12 Months
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.