Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Water, USP/EP Purified, Ricca Chemical

Shop All Ricca Chemical Company Products


Click to view available options
Quantity:
4 L
10 L
20 L
Packaging:
Amber Glass Bottle
Cubitainer
Hard Pack
Natural Poly Bottle
Description
Water, USP/EP Purified
Specifications
Specifications
pH | 5.0 to 7.0 |
Color | Clear, Colorless |
CAS Min % | 98.0 |
CAS Max % | 100.0 |
Conductivity | max. 1 μS/cm |
Endotoxin | max. 0.25 EU/mL at time of manufacture |
Synonym | dihydrogen oxide, dihydrogen monoxide |
IUPAC Name | oxidane |
Molecular Weight (g/mol) | 18.015 |
Percent Purity | 100% |
Show More |
Safety and Handling
ShelfLife | 12 months |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction