Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water, USP Purified, Nonsterile, Ricca Chemical
SDP

Catalog No. 919100055e
Click to view available options
Quantity:
55 gal.
330 gal.
Packaging:
Poly Drum
Tote

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
PubChem CID 962
ChEBI CHEBI:15377
SMILES O

Specifications

Color Colorless
CAS Min % 98.0
CAS Max % 100.0
IUPAC Name oxidane
Molecular Weight (g/mol) 18.015
Percent Purity 100%
Grade USP Purified
Packaging Poly Drum
Quantity 55 gal.
Physical Form Liquid
Chemical Name or Material Water, Non-Sterile
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.