Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ WaterBugs™ Escherichia coli Strain C1

Catalog No. r4751985
Click to view available options
Quantity:
10 Tests/Kit
This item is not returnable. View return policy
This item is not returnable. View return policy

Thermo Scientific™ WaterBugs Escherichia coli Strain C1 is used for water/wastewater quality control testing; microorganisms are supplied in Quanti-Cult format.

Thermo Scientific™ WaterBugs Escherichia coli Strain C1 is used for water/wastewater quality control testing; microorganisms are supplied in Quanti-Cult format.

  • Deliver <50 CFUs in a single inoculum to mimic water samples
  • One test/reconstituted vial

Ordering Alerts:

Supplied with 10 preserved organism vials; 10 rehydration vials.

TRUSTED_SUSTAINABILITY

Specifications

Description Escherichia coli Strain C1
Format Quanti-Cult
Product Type Quality Control Organism
Target Organism Class Gram-negative Rods
Test Type Quantitative QC
Packaging Type 10 Test/Kit
Quantity 10 Tests/Kit

The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.