Learn More
Description
Specifications
Specifications
| Feedwater Source | Tap |
| Flow Rate | 6 L/min. |
| Material | Carbon |
| Pore Size | Fine |
| Purification Method | Activated Carbon |
| Resin Type | Carbon |
| Filter Type | Replacement Prefilter |
| For Use With (Equipment) | WaterPro™ mobile system (Labconco Cat. Nos. 90900-00, -01, 90901-00, -01 and 90902-00, -01) |
| Type | WaterPro Filters |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
