Learn More
Labconco™ WaterPro™ Filters
SDP
Catalog No. 1630718 Shop All Labconco Corporation Products
Change view
Click to view available options
Filter Type:
Activated Carbon
Carbon Prefilter
Deionization Cartridge
Hollow Fiber Final Filter
Organic Adsorption Cartridge
Replacement Prefilter
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Filter Type
16-307-18 Carbon Prefilter
16-307-90A Activated Carbon
16-307-90B Deionization Cartridge
16-307-90C Hollow Fiber Final Filter
16-307-124 Replacement Prefilter
16-307-17 Organic Adsorption Cartridge
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 16-307-18 Supplier Labconco™ Supplier No. 9062200
Add to Cart
Add to Cart
Filters, prefilters and cartridges for Labconco WaterPro PS and RO Water Purification Systems.
Activated Carbon Cartridge aids in removal of contaminants during the creation of Type I ultra pure water. For use with Labconco WaterPro PS water Purification Systems.
Specifications
Feedwater Source Tap
Flow Rate 6 L/min.
Material Carbon
Pore Size Fine
Purification Method Activated Carbon
Resin Type Carbon
Filter Type Carbon Prefilter
For Use With (Equipment) WaterPro™ work station (90000-00, 90001-00, 90002-00 and 90003-00), WaterPro™ reverse osmosis station (90750-00, -02 and 90751-00, -02)
Type WaterPro Filters