Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ WaterPro™ Reverse Osmosis (RO) Systems
SDP

Catalog No. 9075060 Shop All Labconco Corporation Products
Change view
Click to view available options
Electrical Requirements:
115 V 60 Hz
230 V 50/60 Hz
Plug Type:
China/Australia
Domestic
India
North America
Schuko
UK
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Electrical Requirements Plug Type
90-750-60 230 V 50/60 Hz UK
90-750-40 230 V 50/60 Hz North America
90-750-80 230 V 50/60 Hz India
90-750-20 115 V 60 Hz Domestic
90-750-70 230 V 50/60 Hz China/Australia
90-750-30 230 V 50/60 Hz Schuko
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.
Catalog No. 90-750-60 Supplier Labconco™ Supplier No. 9075060
Add to Cart
Add to Cart
Request a Quote
Products in this grouping may have voltages other than the standard US 110/115V. Please review and confirm your selection before completing your order. Some of these products may not be returned.

Three filters (pre, carbon and RO membrane) combined to deliver high-quality reverse osmosis purified water

  • Ideal for laboratory applications such as reagent preparation and glassware rinsing
  • Obtain purified rinse water by connecting to laboratory glassware washers, such as SteamScrubber™ or FlaskScrubber™
  • Timed dispense from 1 to 99.9 min. for unattended operation
  • Temperature compensated conductivity LED monitor displays water quality in microsiemens from 1 to 999
  • Dispense manually from valve or with dispensing gun (sold separately) at rate of 8.7L/min. (tank full) or 1L/min. (tank empty)
  • Storage tank air vent filter removes bacteria, maintaining water quality
  • Automatic flush valve extends RO membrane life by automatically flushing the RO membrane and tank for three minutes after each 12 hour period of inactivity
  • Internal pressure gauge monitors and displays RO-purified water pressure from 0 to 160psi
  • Durable Type 304 stainless steel front panel; powder-coated steel cabinet
  • Large capacity white polypropylene bowls with threaded heads
  • Integral 17L polyethylene storage tank
  • Quiet 1/4 hp motor/rotary vane pump
  • Wall mountable

Certifications

ETL listed.

Specifications

Certifications/Compliance CE, ETL
Capacity (Metric) 17 L
Feed Water Monitoring No
Feedwater Source Tap
Flow Rate 1.1 L/min.
Product Water Type Type 3 to 4
TOC Monitor with UV Intensity Monitoring No
Type Reverse Osmosis System
Display LED
Plug Type UK
Pressure Range 0 to 160 psi
Purification Method Carbon filtration, reverse osmosis
Width (English) 31.1 in.
Width (Metric) 78 cm
Height (English) 30.8 in.
Height (Metric) 78 cm
Depth (English) 7.6 in.
Depth (Metric) 19.3 cm
Weight (English) 90 lb.
Weight (Metric) 40.8 kg
Electrical Requirements 230 V 50/60 Hz
Voltage 230 V
Amperage 6 A
Frequency 50/60 Hz
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.