Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Labconco™ WaterPro™ Reverse Osmosis (RO) Systems

Click to view available options
Electrical Requirements:
115 V 60 Hz
230 V 50/60 Hz
Plug Type:
China/Australia
Domestic
India
North America
Schuko
UK
Description
- Ideal for laboratory applications such as reagent preparation and glassware rinsing
- Obtain purified rinse water by connecting to laboratory glassware washers, such as SteamScrubber™ or FlaskScrubber™
- Timed dispense from 1 to 99.9 min. for unattended operation
- Temperature compensated conductivity LED monitor displays water quality in microsiemens from 1 to 999
- Dispense manually from valve or with dispensing gun (sold separately) at rate of 8.7L/min. (tank full) or 1L/min. (tank empty)
- Storage tank air vent filter removes bacteria, maintaining water quality
- Automatic flush valve extends RO membrane life by automatically flushing the RO membrane and tank for three minutes after each 12 hour period of inactivity
- Internal pressure gauge monitors and displays RO-purified water pressure from 0 to 160psi
- Durable Type 304 stainless steel front panel; powder-coated steel cabinet
- Large capacity white polypropylene bowls with threaded heads
- Integral 17L polyethylene storage tank
- Quiet 1/4 hp motor/rotary vane pump
- Wall mountable
Certifications
ETL listed.
Specifications
Specifications
Certifications/Compliance | CE, ETL |
Capacity (Metric) | 17 L |
Feed Water Monitoring | No |
Feedwater Source | Tap |
Flow Rate | 1.1 L/min. |
Product Water Type | Type 3 to 4 |
TOC Monitor with UV Intensity Monitoring | No |
Type | Reverse Osmosis System |
Display | LED |
Plug Type | North America |
Show More |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction