Learn More
Invitrogen™ WAVE1 Polyclonal Antibody

Description
Immunogen sequence: PRAPHDRRRE WQKLAQGPEL AEDDANLLHK HIEVANGPAS HFETRPQTYV DHMDGSYSLS ALPFSQMSEL LTRAEERVLV RPHEPPPPPP MHGAGDAKPI PTCISSATGL IENRPQSPAT GRTPVFVSPT P Highest antigen sequence indentity to the following orthologs: Rat - 94%, Mouse - 97%.
Specifications
Specifications
| Antigen | WAVE1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | WASF1 |
| Gene Accession No. | Q92558 |
| Gene Alias | AI195380; AI838537; homology of dictyostelium scar 1; KIAA0269; Protein WAVE-1; Scar; SCAR1; Verprolin homology domain-containing protein 1; WAS protein family; WAS protein family member 1; WAS protein family, member 1; WASF1; WASP family 1; WASP family protein member 1; WAVE; WAVE1; WAVE-1; Wiskott-Aldrich syndrome protein family member 1 |
| Gene Symbols | WASF1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.