Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WAVE2 Mouse anti-Human, Mouse, Clone: ZS33-1, MilliporeSigma™
Mouse Monoclonal Antibody
$191.34 - $481.79
Specifications
Antigen | WAVE2 |
---|---|
Clone | ZS33-1 |
Concentration | 0.5 mg/mL |
Applications | Immunofluorescence, Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABT132025
|
MilliporeSigma
MABT132025UL |
25 μL |
Each for $191.34
|
|
|||||
MABT1320100
|
MilliporeSigma
MABT1320100UL |
100 μL |
Each for $481.79
|
|
|||||
Description
WAVE2 Monoclonal antibody specifically detects WAVE2 in Human, Mouse samples. It is validated for Immunofluorescence, Immunoprecipitation, Western Blotting.Specifications
WAVE2 | |
0.5 mg/mL | |
Monoclonal | |
Purified | |
RUO | |
Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. |
ZS33-1 | |
Immunofluorescence, Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse | |
GST-tagged recombinant fragment corresponding to 77 amino acids from the C-terminal region of human Wiskott-Aldrich syndrome protein family member 2 (WAVE-2). | |
Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title