Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WAVE2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560975
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560975 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560975 Supplier Invitrogen™ Supplier No. PA560975
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DSASSPSPSF SEDNLPPPPA EFSYPVDNQR GSGLAGPKRS SVVSPSHPPP AP Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 90%.

WASP (for Wiskott-Aldrich syndrome protein) and N-WASP are downstream effectors of Cdc42 that are implicated in actin polymerization and cytoskeletal organization. The WASP family also includes VASP (vasodilator-stimulated phosphoprotein) and Mena (for mammalian enabled protein), which accumulate at focal adhesions and are also involved in the regulation of the actin cytoskeleton. The WAVE proteins are related to the WASP family proteins and are likewise involved in mediating actin reorganization downstream of the Rho family of small GTPases. The two protein homologs WAVE1 and WAVE2 specifically regulate membrane ruffling by inducing the formation of actin filament clusters in response to GTP binding and activating Rac. The WAVE proteins mediate this actin polymerization by cooperating with the Arp2/3 complex, a nucleation core, and thereby promoting the formation of actin filaments. WAVE1, which is also designated SCAR (for suppressor of cAR), is expressed primarily in the brain, while WAVE2 is widely expressed with the expression highest in peripheral blood leukocytes. WAVE3 forms a multiprotein complex that links receptor kinases and actin. Binding to actin occurs through a C-terminal verprolin homology domain in all family members. The multiprotein complex serves to tranduce signals that involve changes in cell shape, motility or function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WAVE2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.20 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene WASF2
Gene Accession No. Q9Y6W5
Gene Alias AW742646; D4Ertd13e; dJ393P12.2; IMD2; protein WAVE-2; putative Wiskott-Aldrich syndrome protein family member 4; SCAR2; suppressor of cyclic-AMP receptor (WASP-family); verprolin homology domain-containing protein 2; WAS protein family member 2; WAS protein family, member 2; Wasf2; WASF4; WASP family protein member 2; WASP family protein member 4; WASP family Verprolin-homologous protein 2; WAVE2; Wiskott-Aldrich syndrome protein family member 2
Gene Symbols WASF2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human WAVE2. Recombinant protein control fragment (Product #RP-102976).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10163
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.