Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WAVE4 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA599740
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599740 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599740 Supplier Thermo Scientific Supplier No. PA599740

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total WAVE4.

Involved in regulation of actin and microtubule organization. Part of a WAVE complex that activates the Arp2/3 complex. Regulates trichome branch positioning and expansion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WAVE4
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Host Species Rabbit
Immunogen A synthesized peptide derived from human WAVE4, corresponding to a region within C-terminal amino acids.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.