Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ WAY 170523
GREENER_CHOICE

Catalog No. 26331
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-331 1 mg
26-331-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 26-331 Supplier Tocris Bioscience™ Supplier No. 2633/1
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective inhibitor of MMP-13

Potent and selective inhibitor of MMP-13 (IC50 values are 17, 945, > 1000 and > 10000 nM for MMP-13, MMP-9, TACE and MMP-1 respectively).

Chemical Identifiers

CAS 307002-73-9
Molecular Formula C33H31N3O7S

Specifications

Quantity 1 mg
Formula Weight 613.68
Purity >99%
Molecular Weight (g/mol) 613.685
Chemical Name or Material WAY 170523
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.