Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ WAY 181187 oxalate
GREENER_CHOICE

Catalog No. 558910
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
558910 10 mg
558950 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 558910 Supplier Tocris Bioscience™ Supplier No. 5589/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

High affinity and selective 5-HT6 agonist

WAY 181187 oxalate is a high affinity and selective 5-HT6 agonist (Ki = 2.2 nM). Displays 60-fold selectivity over other 5-HT and monoamine receptors. Increases firing rate of 5-HT neurons. Modifies sleep and wakefulness profiles following direct injections into the dorsal raphe nucleus, in vivo. Orally bioavailable.

Specifications

Chemical Name or Material 1-[(6-Chloroimidazo[2,1-b]thiazol-5-yl)sulfonyl]-1H-indole-3-ethanamine oxalate
CAS 1883548-85-3
Quantity 10 mg
Target 5-HT6 Receptor Agonists
Molecular Formula C15H13ClN4O2S2.C2H2O4
Formula Weight 470.91
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.