Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ WAY 213613
GREENER_CHOICE

Catalog No. 265210
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-521-0 10 mg
26-525-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 26-521-0 Supplier Tocris Bioscience™ Supplier No. 2652/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent, non-substrate EAAT2 inhibitor

Potent, non-substrate inhibitor of EAAT2 (GLT-1) that displays > 44-fold selectivity over EAAT1 and EAAT3 (IC50 values are 85, 3787 and 5004 nM for EAAT2, EAAT3 and EAAT1 respectively). Exhibits no activity towards ionotropic and metabotropic glutamate receptors.

Chemical Identifiers

CAS 868359-05-1
Molecular Formula C16H13BrF2N2O4

Specifications

Quantity 10 mg
Formula Weight 415.19
Purity >99%
Molecular Weight (g/mol) 415.191
Chemical Name or Material WAY 213613
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.