Learn More
Description
We are aware of a study in PLOS Biology where WAY-316606 was found to stimulate hair follicle growth. Please note that WAY-316606 from R&D Systems is for laboratory research use only, and that we will only sell to established scientific businesses and institutes.
Specifications
Specifications
| CAS | 1781835-02-6 |
| Quantity | 50 mg |
| Product Line | Tocris |
| Product Type | WAY 316606 hydrochloride |
| Purity | >99% |
| Molecular Weight (g/mol) | 484.94 |
| Inhibitors | WAY 316606 hydrochloride |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

