Learn More
Techcyte WBC/RBC Digital Differential Analysis
Description
- Automates cell locating from a digital image of a blood smear from a glass slide, intended for in-vitro diagnostic use
- Identifies the monolayer and locates, counts and classifies white blood cells, red blood cells, platelets, and attributes of white blood cells from a digital image that has been uploaded to the Techcyte platform
- Intended for blood samples that have been flagged as abnormal by an automated cell counter
Specifications
Specifications
| Product Type | Detection Software |
| For Use With (Application) | Analyzing Data |
Techcyte is CE-IVDD marked in Europe, and for investigational use only in the US.