Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WBP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563396
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563396 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563396 Supplier Invitrogen™ Supplier No. PA563396
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DSGIELCPCP ASGEGEPVKE VRVSATLPDL EDYSPCALPP ESVPQIFPMG LSSSEG Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 73%.

The globular WW domain, named for the conserved tryptophan residues in the protein motif present in various structural and regulatory proteins, is known to play a role in the mediation of protein-protein interactions. This gene encodes a ligand of the WW domain of the Yes kinase-associated protein. Readthrough transcription of the neighboring upstream gene, which encodes INO80 complex subunit B, into this gene generates a non-coding transcript.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WBP1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene WBP1
Gene Accession No. Q96G27
Gene Alias AI255152; WBP1; WBP-1; WW domain binding protein 1; WW domain-binding protein 1
Gene Symbols WBP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human WBP1. Recombinant protein control fragment (Product #RP-101632).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23559
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.