Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ WBP11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561888
Description
Immunogen sequence: KPQITNPKAE ITRFVPTALR VRRENK Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
Posttranslational modification of proteins by the addition of the small protein SUMO.
Specifications
| WBP11 | |
| Polyclonal | |
| Unconjugated | |
| WBP11 | |
| 2510026P17Rik; D6Wsu113e; DKFZp779M1063; hypothetical protein LOC402950; Npw38-binding protein; Npw38-binding protein NpwBP; NpwBP; PPP1R165; protein phosphatase 1, regulatory subunit 165; SH3 domain-binding protein; SH3 domain-binding protein SNP70; SIPP1; SNP70; Splicing factor that interacts with PQBP-1 and PP1; splicing factor, PQBP1 and PP1 interacting; Wbp11; WBP-11; WW domain binding protein 11; WW domain-binding protein 11; zgc:77390 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 51729 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y2W2 | |
| WBP11 | |
| Recombinant protein corresponding to Human WBP11. Recombinant protein control fragment (Product #RP-103046). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction