Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WBP1L Monoclonal Antibody (OPAL1-01), PE
GREENER_CHOICE

Catalog No. PIMA528626
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA528626 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA528626 Supplier Invitrogen™ Supplier No. MA528626
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody recognizes an intracellular epitope of OPAL1 (outcome predictor in acute leukemia 1), a transmembrane adaptor protein expressed mainly by megakaryocytes and platelets.

WBP1L (OPAL1, outcome predictor in acute leukemia 1) is an almost uncharacterized transmembrane adaptor protein expressed mainly by megakaryocytes and platelets. Its expression is enhanced on TEL/AML leukemic cells. The function of OPAL1 is unknown, although the presence of a cytochrome c-like heme-binding site and a transmembrane domain suggested OPAL1 may be involved in the mitochondrial electron transport chain. Its originally reported prognostic impact appears to be treatment dependent.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WBP1L
Applications Flow Cytometry
Classification Monoclonal
Clone OPAL1-01
Concentration 0.1 mg/mL
Conjugate PE
Formulation PBS with 0.2% BSA and 15mM sodium azide; pH 7.4
Gene WBP1L
Gene Accession No. Q9NX94
Gene Alias C10orf26; OPA1L; OPAL1; Outcome predictor in acute leukemia 1; WBP1L; WW domain binding protein 1-like
Gene Symbols WBP1L
Host Species Mouse
Immunogen Recombinant fragment of human OPAL1 (amino acids 152-342).
Purification Method Size-exclusion chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54838
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.