Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ WBP1L Monoclonal Antibody (OPAL1-01)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA528627
Description
This antibody recognizes an intracellular epitope of OPAL1 (outcome predictor in acute leukemia 1), a transmembrane adaptor protein expressed mainly by megakaryocytes and platelets. Western Blot: non-reducing conditions recommended.
WBP1L (OPAL1, outcome predictor in acute leukemia 1) is an almost uncharacterized transmembrane adaptor protein expressed mainly by megakaryocytes and platelets. Its expression is enhanced on TEL/AML leukemic cells. The function of OPAL1 is unknown, although the presence of a cytochrome c-like heme-binding site and a transmembrane domain suggested OPAL1 may be involved in the mitochondrial electron transport chain. Its originally reported prognostic impact appears to be treatment dependent.
Specifications
WBP1L | |
Monoclonal | |
1 mg/mL | |
TBS with 15mM sodium azide; pH 8 | |
Q9NX94 | |
WBP1L | |
Recombinant fragment of human OPAL1 (amino acids 152-342). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
Flow Cytometry, Western Blot | |
OPAL1-01 | |
Unconjugated | |
WBP1L | |
C10orf26; OPA1L; OPAL1; Outcome predictor in acute leukemia 1; WBP1L; WW domain binding protein 1-like | |
Mouse | |
Protein A | |
RUO | |
54838 | |
4°C, do not freeze | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction