Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WBP1L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558361
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558361 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558361 Supplier Invitrogen™ Supplier No. PA558361
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DSGIEVCVCN RGHHDDDLKE FNTLIDDALD GPLDFCDSCH VRPPGDEEEG LCQSSEEQAR EPGHPHLPRP PACLLLNTIN EQDSPNSQ Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

WBP1L (OPAL1, outcome predictor in acute leukemia 1) is an almost uncharacterized transmembrane adaptor protein expressed mainly by megakaryocytes and platelets. Its expression is enhanced on TEL/AML leukemic cells. The function of OPAL1 is unknown, although the presence of a cytochrome c-like heme-binding site and a transmembrane domain suggested OPAL1 may be involved in the mitochondrial electron transport chain. Its originally reported prognostic impact appears to be treatment dependent.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WBP1L
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene WBP1L
Gene Accession No. Q9NX94
Gene Alias C10orf26; OPA1L; OPAL1; Outcome predictor in acute leukemia 1; WBP1L; WW domain binding protein 1-like
Gene Symbols WBP1L
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human WBP1L. Recombinant protein control fragment (Product #RP-97172).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54838
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.