Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WBP2 Monoclonal Antibody (932CT1.3.7)
GREENER_CHOICE

Catalog No. PIMA524757
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524757 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524757 Supplier Invitrogen™ Supplier No. MA524757
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

WBP2 Monoclonal Antibody for Western Blot

The globular WW domain is composed of 38 to 40 semiconserved amino acids shared by proteins of diverse functions including structural, regulatory, and signaling proteins. The domain is involved in mediating protein-protein interactions through the binding of polyproline ligands. WBP2 is a WW domain binding protein, which binds to the WW domain of Yes kinase-associated protein by its PY motifs. The function of this protein has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WBP2
Applications Western Blot
Classification Monoclonal
Clone 932CT1.3.7
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene WBP2
Gene Accession No. P97765, Q969T9
Gene Alias GRAMD6; Wbp2; WBP-2; WW domain binding protein 2; WW domain-binding protein 2
Gene Symbols WBP2
Host Species Mouse
Immunogen recombinant protein between 1-261 amino acids from the C-terminal region of human WBP2.
Purification Method Protein G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22378, 23558
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.