Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WBP2NL Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$254.42 - $728.30
Specifications
| Antigen | WBP2NL |
|---|---|
| Dilution | Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
WBP2NL Polyclonal specifically detects WBP2NL in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| WBP2NL | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| MGC26816, PAWPFLJ26145, postacrosomal sheath WW domain-binding protein, WBP2 N-terminal like, WW domain-binding protein 2-like | |
| WBP2NL | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q6ICG8 | |
| 164684 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: QSHTENRRGALIPNGESLLKRSPNVELSFPQRSEGSNVFSGRKTGTLFLTSYRVIFITSCSISDPMLSFMMPFDLMTNLTVEQPVFAANFIKGTIQAAPYGGWEGQATFKLVFRNGDAIEFAQLMVKAA | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title