Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WBP2NL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561768
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561768 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561768 Supplier Invitrogen™ Supplier No. PA561768
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QSHTENRRGA LIPNGESLLK RSPNVELSFP QRSEGSNVFS GRKTGTLFLT SYRVIFITSC SISDPMLSFM MPFDLMTNLT VEQPVFAANF IKGTIQAAPY GGWEGQATFK LVFRNGDAIE FAQLMVKAA Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 72%.

May play a role in meotic resumption and pronuclear formation, mediated by a WW domain-signaling pathway during fertilization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WBP2NL
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene WBP2NL
Gene Accession No. Q6ICG8
Gene Alias 4930521I23Rik; GRAMD7; PAWP; Postacrosomal sheath WW domain-binding protein; WBP2 N-terminal like; Wbp2nl; WW domain-binding protein 2-like
Gene Symbols WBP2NL
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human WBP2NL. Recombinant protein control fragment (Product #RP-91524).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 164684
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.