Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ WBP2NL Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5106387
Description
Antibody detects endogenous levels of total WBP2NL.
May play a role in meotic resumption and pronuclear formation, mediated by a WW domain-signaling pathway during fertilization.
Specifications
| WBP2NL | |
| Polyclonal | |
| Unconjugated | |
| WBP2NL | |
| 4930521I23Rik; GRAMD7; PAWP; Postacrosomal sheath WW domain-binding protein; WBP2 N-terminal like; Wbp2nl; WW domain-binding protein 2-like | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 164684, 74716 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q6ICG8, Q9D529 | |
| WBP2NL | |
| A synthesized peptide derived from human WBP2NL(Accession Q6ICG8), corresponding to amino acid residues G202-Y252. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction