Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WBP2NL Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501733897 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-3897 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-3897 Supplier Proteintech Group Inc Supplier No. 225871AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

May play a role in meotic resumption and pronuclear formation, mediated by a WW domain-signaling pathway during fertilization.

Specifications

Antigen WBP2NL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene WBP2NL
Gene Accession No. Q6ICG8, Q9D529
Gene Alias PAWP, WBP2 N terminal like, WBP2NL
Gene Symbols Wbp2nl
Host Species Rabbit
Immunogen WBP2NL Fusion Protein Ag18347
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 164684, 74716
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.