Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WBP4 Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP318460R
Description
WBP4 Polyclonal antibody specifically detects WBP4 in Human samples. It is validated for Western BlotSpecifications
WBP4 | |
Polyclonal | |
Western Blot | |
domain-containing binding protein 4, WW domain binding protein 4 (formin binding protein 21) | |
Recognizes region between residues Between Between 125 and 175.. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
DyLight 550 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
11193 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction